Folic acid 98%

Supplier: Ambeed
Warning

Synonyms: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9

A133193-100G A133193-10G A133193-25G A133193-500G A133193-5G A133193-1000G
76767-314EA 30 USD
76767-314 76767-316 76767-318 76767-320 76767-322 77596-792
Folic acid 98%
Folic acid

Caution: for Research Use Only

Formula: C₁₉H₁₉N₇O₆
MW: 441.41 g/mol
MDL Number: MFCD00079305
CAS Number: 59-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR