5-Bromo-2,4-dichloro-6-methylpyrimidine 98%
Supplier: Ambeed
|
Warning
|
A104998-10G
A104998-25G
76765-448EA
14.86
USD
76765-448
77876-803
77876-804
5-Bromo-2,4-dichloro-6-methylpyrimidine 98%
5-Bromo-2,4-dichloro-6-methylpyrimidine
Caution: for Research Use Only
Formula:
C₅H₃BrCl₂N₂ MW: 241.9000 g/mol |
MDL Number:
MFCD00504524 CAS Number: 56745-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...