ActivArmr® 96-002 Goatskin Leather Gloves
Supplier: Ansell Healthcare
RIGLVCVR080 RIGLVCVR090 RIGLVCVR100 RIGLVCVR110 RIGLVCVR120 RIGLVCVR070
76683-128CS 216.34 USD
76683-128 76683-112 76683-138 76683-114 76683-132 76683-120
ActivArmr® 96-002 Goatskin Leather Gloves
Gloves
Goatskin leather protectors, for enhanced low-voltage glove life and improved hand safety. Designed to be worn over Class 0 and 00 electrical safety gloves, these leather protectors can significantly extend wear life.
  • Extend the life of rubber insulated gloves while providing excellent hand protection
  • Provides cut, puncture, and abrasion protection
  • Manufactured to ASTM D120 and EN 60903 standards
  • Suitable to wear with Class 0 and 00 class gloves
ActivArmr® 96-002 protectors are made from premium goatskin leather, enhancing hand safety when conducting electrical work with (or close to) energized components, while offering moderate abrasion, puncture, and cut resistance. The leather formulation also offers excellent dexterity, for genuinely easy-to-use electrical PPE.
Certifications: ASTM F496, ASTM F969, Ecovadis label, EN 388:2016 +A1:2018, EN ISO 21420:2020, ISO 9001.
Packaging: Gloves are individually packaged by pair.