Fluorescent Dyes, M series, Streptavidin Conjugate
Supplier: Abnova
U0279 U0306
10746-284EA 241.54 USD
10746-284 10746-298
Fluorescent Dyes, M series, Streptavidin Conjugate
Protein Linking and Labeling Systems
Fluorescent dye, M series, products are designed to be maximally excited by one of the major light sources equipped in flow cytometers. In comparison with phycobiliprotein tandems, fluorescent dye, M series, possesses better photostability, higher conjugation yields, and little pH sensitivity making it an ideal choice in fluorescence imaging applications.
Recommended usage: Labeling via streptavidin for fluorescence imaging and other fluorescence-based biochemical analysis. For most fluorescent streptavidin applications, a concentration of 1 to 20 g/ml is sufficient. The optimal working dilution should be determined by the end user.