Rhodamine B 97%

Supplier: Ambeed
Danger

A951578-1G A951578-25G A951578-5G A951578-100G
76729-306EA 8 USD
76729-306 76729-308 76729-310 77603-342
Rhodamine B 97%
Rhodamine B

Caution: for Research Use Only

Formula: C₂₈H₃₁ClN₂O₃
MDL Number: MFCD00011931
CAS Number: 81-88-9
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR