Bisacrylamide 97%

Supplier: Ambeed
Danger

Synonyms: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS

A932517-100G A932517-25G A932517-500G A932517-1000G
76729-234EA 7 USD
76729-234 76729-236 76729-238 77552-732
Bisacrylamide 97%
Bisacrylamide

Caution: for Research Use Only

Formula: (H₂C=CHCONH)₂CH₂
MDL Number: MFCD00008625
CAS Number: 110-26-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR