Tripotassium orthophosphate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
Potassium phosphate tribasic, Potassium phosphate
A791587-1000G
A791587-100G
A791587-500G
76728-450EA
18
USD
76728-450
76728-452
76728-454
76728-456
Tripotassium orthophosphate 95%
tri-Potassium orthophosphate
Caution: for Research Use Only
Formula:
K₃O₄P |
MDL Number:
MFCD00036295 CAS Number: 7778-53-2 UN: 3262 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...