Boc-D-3-Pyridylalanine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-β-(3-pyridyl)-D-Ala-OHBoc-D-3-Pal-OHBoc-3-(3-pyridyl)-D-Ala-OH

A790556-250MG
76728-430EA 20.16 USD
76728-430
Boc-D-3-Pyridylalanine 98%
Boc-D-3-Pyridylalanine

Caution: for Research Use Only

Formula: C₁₃H₁₈N₂O₄
MW: 266.297 g/mol
MDL Number: MFCD00076901
CAS Number: 98266-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR