Silver benzoate 98%

Supplier: Ambeed
Warning

Synonyms: Benzoic acid silver (I) salt

A781034-100G A781034-25G A781034-5G
76728-336EA 169 USD
76728-336 76728-338 76728-340
Silver benzoate 98%
Silver benzoate

Caution: for Research Use Only

Formula: C₇H₅AgO₂
MDL Number: MFCD00013030
CAS Number: 532-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR