2,3-Naphthalenedialdehyde (NDA) 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Naphthalene-2,3-dicarboxaldehyde
, Naphthalene-2,3-dicarbaldehyde
, NDA
A770576-250MG
A770576-5G
A770576-1G
A770576-100MG
76728-256EA
85.49
USD
76728-256
76728-254
76728-252
77602-972
2,3-Naphthalenedialdehyde (NDA) 99%
2,3-Naphthalenedialdehyde (NDA)
Caution: for Research Use Only
Formula:
C₁₂H₈O₂ MW: 184.19 g/mol |
MDL Number:
MFCD00016618 CAS Number: 7149-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...