n-Pentadecane 98%

Supplier: Ambeed
Danger

A735739-25G A735739-100G A735739-500G
76728-008EA 22 USD
76728-008 77880-502 77880-503
n-Pentadecane 98%
n-Pentadecane

Caution: for Research Use Only

Formula: H₃C(CH₂)₁₃CH₃
MDL Number: MFCD00008990
CAS Number: 629-62-9
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR