N-Cyclohexylformamide 98%

Supplier: Ambeed
Danger

Synonyms: N-cyclohexylformamide

A728657-25G A728657-5G
76727-944EA 45 USD
76727-944 76727-946
N-Cyclohexylformamide 98%
N-Cyclohexylformamide

Caution: for Research Use Only

Formula: C₇H₁₃NO
MW: 127.19 g/mol
MDL Number: MFCD00003828
CAS Number: 766-93-8
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR