Sodium thiosulfate 98%

Supplier: Ambeed
Warning

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

A716445-100G A716445-500G
76727-838EA 5 USD
76727-838 76727-840
Sodium thiosulfate 98%
Sodium thiosulphate

Caution: for Research Use Only

Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
MDL Number: MFCD00003499
CAS Number: 7772-98-7
Merck Index: 12,08844

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR