n-Heptadecane 99%

Supplier: Ambeed
Danger

A705883-25G A705883-500G A705883-5G
76727-734EA 17 USD
76727-734 77669-900 77880-465
n-Heptadecane 99%
n-Heptadecane

Caution: for Research Use Only

Formula: H₃C(CH₂)₁₅CH₃
MDL Number: MFCD00009002
CAS Number: 629-78-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR