Pentafluorobenzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: α-Bromo-2,3,4,5,6-pentafluorotoluene2,3,4,5,6-Pentafluorobenzyl bromide

A682905-5G A682905-25G A682905-1G
77552-078EA 14.42 USD
77552-078 77552-080 76727-588
Pentafluorobenzyl bromide 97%
Pentafluorobenzyl bromide

Caution: for Research Use Only

Formula: (C₆F₅)CH₂Br
MW: 260.99 g/mol
MDL Number: MFCD00000299
CAS Number: 1765-40-8
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR