N-(Hydroxymethyl)-2,2,2-trifluoroacetamide 95%

Supplier: Ambeed
Warning

Synonyms: 2,2,2-Trifluoro-N-(hydroxymethyl)acetamide, N-(Hydroxymethyl)trifluoroacetamide

A666312-100G A666312-10G A666312-1G A666312-5G A666312-25G
76727-468EA 247 USD
76727-468 76727-470 76727-472 76727-474 77669-816
N-(Hydroxymethyl)-2,2,2-trifluoroacetamide 95%
N-(Hydroxymethyl)-2,2,2-trifluoroacetamide

Caution: for Research Use Only

Formula: C₃H₄F₃NO₂
MW: 143.06 g/mol
MDL Number: MFCD00014416
CAS Number: 50667-69-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR