Methyl orange 98%

Supplier: Ambeed
Danger

Synonyms: 4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulphonic acid, sodium saltMethyl orange sodium saltHelianthine4-((4-(Dimethylamino)phenyl)diazenyl)benzenesulfonic acid, sodium salt

A630312-25G
76727-146EA 6.37 USD
76727-146
Methyl orange 98%
Methyl orange

Caution: for Research Use Only

Formula: C₁₄H₁₄N₃NaO₃S
MW: 327.34 g/mol
MDL Number: MFCD00007502
CAS Number: 547-58-0
UN: 3143
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR