Sodium L(+)-ascorbate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
A587658-100G
A587658-100MG
A587658-10G
A587658-25G
A587658-500G
A587658-1000G
76726-796EA
11.67
USD
76726-796
76726-798
76726-800
76726-802
76726-804
77602-456
Sodium L(+)-ascorbate 98%
Sodium L(+)-ascorbate
Caution: for Research Use Only
Formula:
C₆H₇NaO₆ MW: 198.11 g/mol |
MDL Number:
MFCD00082340 CAS Number: 134-03-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...