(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Aminomethy-4-(4-fluorobenzyl)morpholine
A544915-100G
A544915-10G
A544915-1G
A544915-250MG
A544915-25G
A544915-5G
76726-406EA
153
USD
76726-406
76726-408
76726-410
76726-412
76726-414
77669-408
(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine 98%
(4-(4-Fluorobenzyl)morpholin-2-yl)methanamine
Caution: for Research Use Only
Formula:
C₁₂H₁₇FN₂O MW: 224.28 g/mol |
MDL Number:
MFCD06658145 CAS Number: 112914-13-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...