Nefiracetam 97%

Supplier: Ambeed
Warning

Synonyms: N-(2,6-Dimethylphenyl)-2-(2-oxopyrrolidin-1-yl)acetamide

A517196-100MG A517196-1G A517196-1MG A517196-250MG A517196-25G A517196-50MG A517196-5G
76726-174EA 28 USD
76726-174 76726-176 76726-178 76726-180 76726-182 76726-184 76726-186
Nefiracetam 97%
Nefiracetam

Caution: for Research Use Only

Formula: C₁₄H₁₈N₂O₂
MW: 246.31 g/mol
MDL Number: MFCD00209882
CAS Number: 77191-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR