Oxalic acid 95%

Supplier: Ambeed
Danger

Synonyms: Ethanedioic acid

A473398-100G A473398-500G A473398-1000G
76725-810EA 10 USD
76725-810 76725-812 77568-932
Oxalic acid 95%
Oxalic acid

Caution: For research use only.

Formula: C₂H₂O₄
MDL Number: MFCD00002573
CAS Number: 144-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR