n-Decyl-β-D-maltoside 98%

Supplier: Ambeed
Warning

A466195-5G A466195-1G A466195-250MG A466195-25G
76725-746EA 146 USD
76725-746 77602-036 77602-038 77669-098
n-Decyl-β-D-maltoside 98%
n-Decyl-β-D-maltoside

Caution: for Research Use Only

Formula: C₂₂H₄₂O₁₁
MDL Number: MFCD00061624
CAS Number: 82494-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR