Riboflavine (vitamin B2) 98%

Supplier: Ambeed
Warning

A454767-1000G A454767-100G A454767-10G A454767-1G A454767-500G A454767-5G A454767-25G
76725-604EA 199 USD
76725-604 76725-606 76725-608 76725-610 76725-612 76725-614 76842-738
Riboflavine (vitamin B2) 98%
Riboflavine (Vitamin B2)

Caution: for Research Use Only

Formula: C₁₇H₂₀N₄O₆
MDL Number: MFCD00005022
CAS Number: 83-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR