N-Isopropylcyclohexylamine 97%

Supplier: Ambeed
Danger

A422816-1G A422816-5G
76725-258EA 17.01 USD
76725-258 77668-892
N-Isopropylcyclohexylamine 97%
N-Isopropylcyclohexylamine

Caution: for Research Use Only

Formula: C₉H₁₉N
MDL Number: MFCD00003831
CAS Number: 1195-42-2
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR