Sodium thiosulfate pentahydrate 95%
Supplier: Ambeed
|
Warning
|
A355130-1000G
A355130-100G
A355130-500G
76724-516EA
11
USD
76724-516
76724-518
76724-520
Sodium thiosulfate pentahydrate 95%
Sodium thiosulphate pentahydrate
Caution: for Research Use Only
Formula:
Na₂S₂O₃.5H₂O MW: 248.19 g/mol Melting Pt: 48 |
MDL Number:
MFCD00149186 CAS Number: 10102-17-7 Merck Index: 12,08844 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...