(S)-2-(Dimethylamino)-3-methylbutanoic acid 97%
Supplier: Ambeed
|
Warning
|
A341677-1G
A341677-5G
A341677-250MG
76724-348EA
37
USD
76724-348
77593-996
77879-734
(S)-2-(Dimethylamino)-3-methylbutanoic acid 97%
(S)-2-(Dimethylamino)-3-methylbutanoic acid
Caution: for Research Use Only
Formula:
C₇H₁₅NO₂ |
MDL Number:
MFCD08059008 CAS Number: 2812-32-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...