Nɑ-Acetyl-L-glutamine 97%

Supplier: Ambeed
Warning

A324419-100MG A324419-10G A324419-25G A324419-100G A324419-500G
76724-016EA 5 USD
76724-016 76724-018 76724-020 77668-448 77668-450
Nɑ-Acetyl-L-glutamine 97%
Nalpha-Acetyl-L-glutamine

Caution: for Research Use Only

Formula: C₇H₁₂N₂O₄
MDL Number: MFCD00038159
CAS Number: 2490-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR