O-tert-Butyl-L-tyrosine 98%

Supplier: Ambeed
Warning

Synonyms: H-Tyr(tBu)-OH

A280125-100G A280125-10G A280125-1G A280125-250MG A280125-25G A280125-5G A280125-500G
76723-260EA 143 USD
76723-260 76723-262 76723-264 76723-266 76723-268 76723-270 77879-526
O-tert-Butyl-L-tyrosine 98%
O-tert-Butyl-L-tyrosine

Caution: for Research Use Only

Formula: C₁₃H₁₉NO₃
MDL Number: MFCD00037202
CAS Number: 18822-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR