Nocodazole 98%

Supplier: Ambeed
Warning

Synonyms: Methyl (6-(thiophene-2-carbonyl)-1H-benzo[d]imidazol-2-yl)carbamate

A260772-1MG A260772-100MG A260772-50MG
76722-800EA 12.74 USD
76722-800 76722-802
Nocodazole 98%
Nocodazole

Caution: for Research Use Only

Formula: C₁₄H₁₁N₃O₃S
MW: 301.32 g/mol
MDL Number: MFCD00005588
CAS Number: 31430-18-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR