9-Ethylcarbazole 99%

Supplier: Ambeed
Warning

A256020-100G A256020-500G A256020-25G
76722-668EA 23 USD
76722-668 76722-670 77091-558
9-Ethylcarbazole 99%
9-Ethylcarbazole

Caution: for Research Use Only

Formula: C₁₄H₁₃N
MDL Number: MFCD00004967
CAS Number: 86-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR