2-Amino-2'-chloro-5-nitrobenzophenone 97%

Supplier: Ambeed
Warning

Synonyms: Clonazepam Related Compound B, 2-Amino-5-nitro-2’-chlorobenzophenone, 2-Amino-2'-chloro-5-nitrobenzophenone, (2-Amino-5-nitrophenyl)(2-chlorophenyl)methanone

A217770-100G A217770-10G A217770-25G A217770-5G A217770-500G
76721-862EA 167 USD
76721-862 76721-864 76721-866 76721-868 77600-754
2-Amino-2'-chloro-5-nitrobenzophenone 97%
2-Amino-2'-chloro-5-nitrobenzophenone

Caution: for Research Use Only

Formula: C₁₃H₉ClN₂O₃
MDL Number: MFCD00792453
CAS Number: 2011-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR