Platinum(II) chloride 98%

Supplier: Ambeed
Danger

A201173-100MG A201173-10G A201173-250MG A201173-5G A201173-1G A201173-25G
76721-458EA 15 USD
76721-458 76721-460 76721-462 76721-464 77550-290 77667-670
Platinum(II) chloride 98%
Platinum (II) chloride

Caution: for Research Use Only

Formula: Cl₂Pt
MDL Number: MFCD00011181
CAS Number: 10025-65-7
UN: 3260
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR