Propylene carbonate 97%

Supplier: Ambeed
Warning

Synonyms: Propylene glycol carbonate, 4-Methyl-1,3-dioxolan-2-one, Carbonic acid propylene glycol ester

A184527-1000G A184527-100G A184527-500G
76720-986EA 23 USD
76720-986 76720-988 76720-990
Propylene carbonate 97%
Propylene carbonate

Caution: for Research Use Only

Formula: C₄H₆O₃
MDL Number: MFCD00005385
CAS Number: 108-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR