n-Heptacosane 98%

Supplier: Ambeed
Warning

Synonyms: Heptacosane

A173057-1G A173057-250MG A173057-100MG
76720-570EA 32.89 USD
76720-570 76720-572
n-Heptacosane 98%
n-Heptacosane

Caution: for Research Use Only

Formula: H₃C(CH₂)₂₅CH₃
MW: 380.74 g/mol
MDL Number: MFCD00009862
CAS Number: 593-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR