Methyl salicylate 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Wintergreen Oil, Methyl 2-hydroxybenzoate
A171195-1000G
A171195-100G
A171195-10G
A171195-25G
A171195-500G
76720-518EA
18
USD
76720-518
76720-520
76720-522
76720-524
76720-526
Methyl salicylate 95%
Methyl salicylate
Caution: for Research Use Only
Formula:
2-(HO)C₆H₄CO₂CH₃ MW: 152.15 g/mol Boiling Pt: 432 Flash Pt: 99 |
MDL Number:
MFCD00002214 CAS Number: 119-36-8 Merck Index: 13,06143 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...