10H-Phenothiazine-10-propanesulfonic acid sodium salt 96%

Supplier: Ambeed
Warning

A169900-100G A169900-1G A169900-250MG A169900-25G A169900-5G
76720-458EA 1783 USD
76720-458 76720-460 76720-462 76720-464 76720-466
10H-Phenothiazine-10-propanesulfonic acid sodium salt 96%
10H-Phenothiazine-10-propanesulfonic acid sodium salt

Caution: for Research Use Only

Formula: C₁₅H₁₄NNaO₃S₂
MDL Number: MFCD08275674
CAS Number: 101199-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR