myo-Inositol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Inositol
, rel-(1R,2r,3S,4R,5s,6S)-Cyclohexane-1,2,3,4,5,6-hexaol
, i-Inositol
, D-Myo-Inositol
A169757-100G
A169757-5G
A169757-1000G
A169757-500G
A169757-25G
76720-448EA
9.27
USD
76720-448
76720-446
76720-454
76720-452
76720-450
myo-Inositol 98%
myo-Inositol
Caution: for Research Use Only
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol |
MDL Number:
MFCD00077932 CAS Number: 87-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...