Phloridzin 98%

Supplier: Ambeed
Warning

A163609-100G A163609-10G A163609-1G A163609-250MG A163609-25G A163609-5G
76720-228EA 161 USD
76720-228 76720-230 76720-232 76720-234 76720-236 76720-238
Phloridzin 98%
Phloridzin

Caution: for Research Use Only

Formula: C21H24O10
MDL Number: MFCD00006591
CAS Number: 60-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR