Sodium oxamate 95%

Supplier: Ambeed
Warning

Synonyms: Oxamic acid sodium salt, Sodium 2-amino-2-oxoacetate

A154629-100G A154629-25G A154629-5G
76719-884EA 204 USD
76719-884 76719-886 76719-888 76719-890
Sodium oxamate 95%
Sodium oxamate

Caution: for Research Use Only

Formula: C₂H₂NNaO₃
MDL Number: MFCD00044553
CAS Number: 565-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR