Methylene blue 98%

Supplier: Ambeed
Warning

Synonyms: Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue

A151225-100G A151225-1G A151225-25G A151225-25MG A151225-500G A151225-5G
76719-730EA 22 USD
76719-730 76719-732 76719-734 76719-736 76719-738 76719-740
Methylene blue 98%
Methylene blue

Caution: for Research Use Only

Formula: C₁₆H₁₈ClN₃S
MW: 319.86 g/mol
Melting Pt: 180
MDL Number: MFCD00150008
CAS Number: 61-73-4
Merck Index: 12,06137

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR