L(+)-Sodium lactate 90%

Supplier: Ambeed
Warning

Synonyms: (S)-(+)-2-Hydroxypropionic acid sodium salt(S)-(+)-Lactic acid sodium salt(S)-(+)-2-Hydroxypropanoic acid sodium saltL(+)-Lactic acid sodium saltSodium (S)-2-hydroxypropanoateSodium-(S)-lactate

A149141-100G A149141-500G A149141-1000G
76719-672EA 31.83 USD
76719-672 76719-674 76719-676
L(+)-Sodium lactate 90%
L(+)-Sodium lactate

Caution: for Research Use Only

Formula: C₃H₅NaO₃
MW: 112.06 g/mol
MDL Number: MFCD00066576
CAS Number: 867-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR