MOPSO (3-(4-morpholino)-2-hydroxypropanesulfonic acid) 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Hydroxy-3-morpholinopropane-1-sulphonic acid, 2-Hydroxy-3-morpholinopropane-1-sulfonic acid
A136758-1000G
A136758-100G
A136758-25G
A136758-500G
76719-214EA
77
USD
76719-214
76719-216
76719-218
76719-220
76719-222
MOPSO (3-(4-morpholino)-2-hydroxypropanesulfonic acid) 99%
MOPSO (3-(4-Morpholino)-2-hydroxypropanesulphonic acid)
Caution: for Research Use Only
Formula:
C₇H₁₅NO₅S |
MDL Number:
MFCD00009629 CAS Number: 68399-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...