DL-Sodium lactate 60% in water

Supplier: Ambeed
Warning

A128732-100G A128732-500G A128732-1000G
76718-872EA 7 USD
76718-872 76718-874 77599-878
DL-Sodium lactate 60% in water
DL-Sodium lactate

Caution: for Research Use Only

Formula: H₃CCH(OH)COONa
MDL Number: MFCD00065400
CAS Number: 72-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR