Spermidine 98% (by GC)

Supplier: Ambeed
Danger

Synonyms: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine

A128137-1G A128137-250MG A128137-25G A128137-5G
76718-836EA 24 USD
76718-836 76718-838 76718-840 76718-842
Spermidine 98% (by GC)
Spermidine

Caution: for Research Use Only

Formula: C₇H₁₉N₃
MW: 145.25 g/mol
Boiling Pt: 246
MDL Number: MFCD00008229
CAS Number: 124-20-9
UN: 2735
ADR: 8 Ⅲ
Merck Index: 12,08893

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR