10-Hydroxybenzo[h]quinoline 98%

Supplier: Ambeed
Warning

A119546-1G A119546-25G A119546-5G A119546-100G
76718-462EA 9 USD
76718-462 76718-464 77566-206 77566-208 77871-515
10-Hydroxybenzo[h]quinoline 98%
10-Hydroxybenzo[h]quinoline

Caution: For research use only.

Formula: C₁₃H₉NO
MDL Number: MFCD00142350
CAS Number: 33155-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR