Nitrofurantoin 98+%
Supplier: Ambeed
|
Danger
|
Synonyms:
1-[(5-Nitrofurfurylidene)amino]hydantoin
A109585-100G
A109585-1G
A109585-25G
A109585-500G
A109585-5G
A109585-1MG
76718-010EA
47
USD
76718-010
76718-012
76718-014
76718-016
76718-018
76718-020
77666-596
Nitrofurantoin 98+%
Nitrofurantoin
Caution: for Research Use Only
Formula:
C₈H₆N₄O₅ MW: 238.16 g/mol |
MDL Number:
MFCD00003224 CAS Number: 67-20-9 UN: 2811 ADR: 6.1 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...