N,N'-Bis(tert-butyl)ethylenediamine 98%

Supplier: Ambeed
Danger

A107184-25G A107184-5G
76717-894EA 11 USD
76717-894 76717-896
N,N'-Bis(tert-butyl)ethylenediamine 98%
N,N'-Bis(tert-butyl)ethylenediamine

Caution: for Research Use Only

Formula: C₁₀H₂₄N₂
MDL Number: MFCD00014996
CAS Number: 4062-60-6
UN: 2735
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR