HALO2 Wireless Field pH Tester, PEI Body
Supplier: Hanna
HI9810422 HI9810422
470349-838EA 166.79 USD
CA470349-838 470349-838
HALO2 Wireless Field pH Tester, PEI Body
Electrochemistry Meters pH and Multi-Parameter Meters
Our HALO2 Wireless Field pH Tester has a durable PEI body and spherical sensing bulb, perfect for use in the field.
  • Open-source Bluetooth allows easy integration into current data logging systems
  • IP 65 water resistant protection
PEI is suitable for a wide range of applications and excels in field measurements due to its durability. The shielding around the spherical glass tip minimizes breakage due to accidental bumping or dropping of the electrode. PEI is a high-quality plastic that is chemically resistant to many aggressive chemicals.
Cleaning and Disinfection Solution-Your HALO2 pH Tester comes with cleaning solution that has been engineered to keep your electrode free from grease and debris. This concentrated combination of detergents cleans your meter quickly and effectively so that your meter stays in optimum condition.
Easy Calibration-HALO2 can be placed right into our calibration buffer sachets for easy calibration. Using a Hanna calibration sachet ensures your buffer is always fresh.
Storage Solution-Proper storage of your electrode will ensure reliable measurements and a long electrode life. Our storage solution is specially formulated to keep the pH glass hydrated while minimizing organic growth.
Ordering information: If pH probe dies, will have to repurchase product. pH calibration solutions, storage solutions, and cleaning solutions sold seperately. Contact your sales representative for more information.