Glass Delivery Tubes
Supplier: EISCO SCIENTIFIC LLC
CH0749
470344-566EA 5.29 USD
470344-566
Glass Delivery Tubes
Tubes Gas Inlet Tubes
This set of six borosilicate glass delivery tubes offers durability and chemical resistance, ensuring efficient and precise liquid transfer in laboratory applications.
  • Set of six uniquely shaped glass delivery tubes
  • Thick borosilicate glass 1 mm
  • Internal diameter is 4 mm
  • External diameter is 6 mm
  • See pictures for tube lengths
Set of six variably shaped glass delivery tubes made of borosilicate glass. Interior diameter of each tube is 4 mm and exterior diameter is 6 mm making for a 1 mm thick glass wall. This economically priced, diverse assortment of shapes works for a variety of lab configurations. Autoclavable, with superior chemical durability. Low temperature gradient- can withstand high temperatures and thermal shocks. Eisco borosilicate glass has the following heat tolerances: Strain point 515 °C, annealing point 565 °C, softening point 820 °C.