Synthware High Vacuum Valves, PTFE, O-Ring, Three-Way
Supplier: Kemtech America
S580008 S580012
60004-004EA 270.45 USD
60004-004 60004-006
Synthware High Vacuum Valves, PTFE, O-Ring, Three-Way
Valves Vacuum Valves
Valves feature two 9 mm O.D. sidearms sealed to the barrel at a 90° angle to one another.
  • Heavy wall construction
  • 33 expansion Borosilicate glass
The non-rotating PTFE plug has three O-Rings. The tip O-Ring makes the primary seal on the precision formed taper seat.